Skip to Content
Merck
All Photos(2)

Key Documents

707503

Sigma-Aldrich

Small gap fullerenes

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

granular

mol wt

average Mw ~1,000

composition

fullerenes, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>400 °C

solubility

organic solvents: soluble

greener alternative category

SMILES string

C=1=C2c3c4C=1c5c6c7c8c9c%10c%11c%12c%13C%14=[C]%10=C%15c%16c%17c%14c%18c%13c%19c%20c%12c%21c%11c(c%22c%23c%24c(C2=[C]7%15%22)c%25c3c%26c%27c4c%28c5c(c6%16)c%29c%17c%30c%18c%31c%19c%32c%20c(c%21%23)c%33c%24c%34c%25c%26c%35c%36c%27c%28c%29c%37c%30c%31c%38c%32c%33c%34c%35c%38c%36%37)c89

General description

Insoluble small gap fullerenes can be extracted into organic solvents by simple chemical redox process.[1]
Small gap fullerenes are higher order fullerenes with low HOMO − LUMO gaps. These allotropes of carbon are also known as metallofullerenes. They are kinetically unstable and are considered as insoluble small band gap structures. They can be extracted into organic solvents by chemical redox processes.[2][3]
Small gap fullerenes, also known as giant fullerenes, possess small energy differences between the valence band and the conduction band or between the filled and unfilled energy states of individual fullerene molecules.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Small gap fullerenes can be used as doping agents on polyaniline (PANI) for organic electronics applications.[5]

Legal Information

Product of TDA Research, Inc. and US Patents 6,517,799 and 6,303,016.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical redox recovery of giant, small-gap and other fullerenes
Raebiger JW, et al.
Carbon, 49(1), 37-46 (2011)
Optical, structural and electrical properties of polyaniline systems doped with C60 and small gap C60 fullerenes
Politakos N, et al.
Materials Chemistry and Physics, 142(1), 387-394 (2013)
Chemical redox recovery of giant, small-gap and other fullerenes
Raebiger, J.W.; Alford, J.M.; Bolskar, R.D.; Diener, M.D.
Carbon, 49, 37-46 (2011)
Small-bandgap endohedral metallofullerenes in high yield and purity.
Stevenson S, et al.
Nature, 401(6748), 55-55 (1999)
Isolation and properties of small-bandgap fullerenes.
Diener MD and Alford JM
Nature, 393(6686), 668-668 (1998)

Articles

Find various photovoltaic and bioscience-based applications of fullerenes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service