Skip to Content
Merck
All Photos(1)

Key Documents

694991

Sigma-Aldrich

5-Hexenylboronic acid

Synonym(s):

Hex-5-enylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13BO2
CAS Number:
Molecular Weight:
127.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

67-73 °C

functional group

allyl

storage temp.

2-8°C

SMILES string

OB(O)CCCCC=C

InChI

1S/C6H13BO2/c1-2-3-4-5-6-7(8)9/h2,8-9H,1,3-6H2

InChI key

JXFKAWIGNACKQN-UHFFFAOYSA-N

General description

may contain varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service