675547
1,2-Bis[(2S,5S)-2,5-dimethylphospholano]ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
About This Item
Recommended Products
form
solid
mp
152.2-156.3 °C
SMILES string
[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.C[C@H]2CC[C@H](C)P2CCP3[C@@H](C)CC[C@@H]3C
InChI
1S/C14H28P2.C8H12.BF4.Rh/c1-11-5-6-12(2)15(11)9-10-16-13(3)7-8-14(16)4;1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h11-14H,5-10H2,1-4H3;1-2,7-8H,3-6H2;;/q;;-1;+1/b;2-1-,8-7-;;/t11-,12-,13-,14-;;;/m0.../s1
InChI key
CVUPEPGZCZHSOF-ASDDUFFSSA-N
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service