Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655813

Sigma-Aldrich

6-(1,3-Benzodioxol-5-yl)-2-pyridinecarboxaldehyde

97%

Synonym(s):

6-(Benzo[1,3]dioxol-5-yl)pyridine-2-carbaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$143.00
5 G
$520.00

Vendor SKU: LD-0881-1G

$143.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$143.00
5 G
$520.00

About This Item

Empirical Formula (Hill Notation):
C13H9NO3
CAS Number:
Molecular Weight:
227.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Vendor SKU: LD-0881-1G

$143.00


Ships in 7 days - from Aldrich Partner.

Assay

97%

mp

117-122 °C

SMILES string

[H]C(=O)c1cccc(n1)-c2ccc3OCOc3c2

InChI

1S/C13H9NO3/c15-7-10-2-1-3-11(14-10)9-4-5-12-13(6-9)17-8-16-12/h1-7H,8H2

InChI key

XITSUEJTFNGOIW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service