Skip to Content
Merck
All Photos(1)

Documents

594881

Sigma-Aldrich

2,6-Dimethylphenyl isocyanide

96%

Synonym(s):

vic.-m-Xylyl isocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NC
CAS Number:
Molecular Weight:
131.17
Beilstein:
3541909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

72-76 °C

SMILES string

Cc1cccc(C)c1[N+]#[C-]

InChI

1S/C9H9N/c1-7-5-4-6-8(2)9(7)10-3/h4-6H,1-2H3

InChI key

DNJLFZHMJDSJFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dimethylphenyl isocyanide may be used in the synthesis of N-(2,6-dimethylphenyl)diphenyliminocyclobutenone via reaction with diphenylcyclopropenone catalyzed by triphenylphosphine. 2,6-dimethylphenyl isocyanide can also undergo (4+1) cycloaddition reaction with certain α,β-unsaturated ketones in the presence of GaCl3 catalyst to give the corresponding unsaturated lactones.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ugi I
Isonitrile Chemistry (2012)
Knipe.C.A
Organic Reaction Mechanisms, 124 (2008)
M A Wood et al.
The Biochemical journal, 247(3), 675-678 (1987-11-01)
A series of aromatic isonitriles were synthesized and their binding to sheep haemoglobin and horse heart myoglobin was investigated. The disubstituted ligands 2,6-dimethylphenylisonitrile and 2,6-diethylphenylisonitrile were found to bind to horse-heart myoglobin with affinities ranging from 500 to 5000 times

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service