Skip to Content
Merck
All Photos(1)

Documents

560197

Sigma-Aldrich

3-Bromo-4-methoxyphenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)(CH2CH2NH2)
CAS Number:
Molecular Weight:
230.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D >1.5800 (lit.)

density

1.389 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CCN)cc1Br

InChI

1S/C9H12BrNO/c1-12-9-3-2-7(4-5-11)6-8(9)10/h2-3,6H,4-5,11H2,1H3

InChI key

JHVALSRTUOVNLL-UHFFFAOYSA-N

General description

3-Bromo-4-methoxyphenethylamine can be synthesized by reacting N-benzoyl-3-bromo-4-methoxyphenethylamine, acetic acid and HCl in the presence of N2.

Application

3-Bromo-4-methoxyphenethylamine (3-Bromo-4-methoxy-β-phenethylamine) may be used as a starting reagent in the synthesis of 3-bromo-4-methoxy-5-nitro-β-phenethylamine. It may also be used to synthesize 4-bromo-N-(3-bromo-4-methoxyphenethyl)-1H-pyrrole-2-carboxamide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Aminoisoquinolines and Related Compounds. X. A Modified Synthesis of dl-Cularine.
Ishiwata S, et al.
Chemical & Pharmaceutical Bulletin, 18(9), 1850-1855 (1970)
Total synthesis and biological evaluation of the marine bromopyrrole alkaloid dispyrin: elucidation of discrete molecular targets with therapeutic potential.
Kennedy JP, et al.
Journal of Natural Products, 1783-1786 (2008)
Kumar Saurav et al.
Marine drugs, 18(2) (2020-02-26)
Marine sponges, a well-documented prolific source of natural products, harbor highly diverse microbial communities. Their extracts were previously shown to contain quorum sensing (QS) signal molecules of the N-acyl homoserine lactone (AHL) type, known to orchestrate bacterial gene regulation. Some
Syntheses of the marine metabolites verongamine, hemibastadin-2, and aerothionin using the cyano ylide coupling methodology.
Wasserman HH and Wang J.
The Journal of Organic Chemistry, 63(16), 5581-5586 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service