Skip to Content
Merck
All Photos(1)

Documents

557560

Sigma-Aldrich

Ethyl phenyl sulfone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2CH3
CAS Number:
Molecular Weight:
170.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

35-40 °C (lit.)

SMILES string

CCS(=O)(=O)c1ccccc1

InChI

1S/C8H10O2S/c1-2-11(9,10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

VBQUDDWATQWCPP-UHFFFAOYSA-N

Gene Information

General description

Density of ethyl phenyl sulfone is 1.1410g/cm3 at 20°C.

Application

Ethyl phenyl sulfone may be used in the preparation of (±)-α-multistriatin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly stereoselective synthesis of (?)-a-multistriatin.
Bartlett PA and Myerson J.
The Journal of Organic Chemistry, 44(10), 1625-1627 (1979)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 197-197 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service