Skip to Content
Merck
All Photos(1)

Documents

551171

Sigma-Aldrich

1-(4-Methylphenyl)-1-propanol

97%

Synonym(s):

1-(p-Tolyl)-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(OH)CH2CH3
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5180 (lit.)

bp

234-235 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

SMILES string

CCC(O)c1ccc(C)cc1

InChI

1S/C10H14O/c1-3-10(11)9-6-4-8(2)5-7-9/h4-7,10-11H,3H2,1-2H3

InChI key

ZBDUWPZEVMGAMD-UHFFFAOYSA-N

General description

1-(4-Methylphenyl)-1-propanol is a secondary alcohol derivative that can be prepared by the reduction of 4′-methylpropiophenone. Its racemic form can be resolved by oxidative kinetic resolution in the presence of chiral polymeric Mn(III) salen complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Easily recyclable chiral polymeric Mn (III) salen complexes for oxidative kinetic resolution of racemic secondary alcohols.
Kureshy RI, et al.
Chirality, 19(5), 352-357 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service