Skip to Content
Merck
All Photos(1)

Key Documents

469084

Sigma-Aldrich

DAB-Am-32, Polypropylenimine dotriacontaamine Dendrimer, Generation 4.0

volume 5,054 Å3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2N[(CH2)3N[(CH2)3N[(CH2)3N[(CH2)3NH2]2]2]2]2]2
CAS Number:
Molecular Weight:
3513.81
MDL number:
UNSPSC Code:
12162002

mol wt

calculated mol wt 3,514

volume

5,054 Å3

refractive index

n20/D 1.518 (lit.)

viscosity

2.5 poise(50 °C)(lit.)

transition temp

Tg −87 °C

solubility

methanol and water: soluble

density

1.01 g/mL at 25 °C (lit.)

SMILES string

NCCCN(CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN(CCCCN(CCCN(CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN)CCCN(CCCN(CCCN)CCCN)CCCN(CCCN)CCCN

Physical form

Contains 32 primary amino groups.

Legal Information

DSM product

comparable product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service