Skip to Content
Merck
All Photos(1)

Documents

410594

Sigma-Aldrich

2-Chloroethyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2NCS
CAS Number:
Molecular Weight:
121.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.556 (lit.)

bp

80 °C/13 mmHg (lit.)

density

1.265 g/mL at 25 °C (lit.)

SMILES string

ClCCN=C=S

InChI

1S/C3H4ClNS/c4-1-2-5-3-6/h1-2H2

InChI key

ZUWFBQUHBOUPFK-UHFFFAOYSA-N

General description

2-Chloroethyl isothiocyanate is a chloroalkyl isothiocyanate derivative.

Application

2-Chloroethyl isothiocyanate may be used in the following studies:
  • To investigate the electrochemical properties of sulforaphane (SFR) by cyclic voltammetry and electrochemical quartz crystal microbalance (EQCM) methods.
  • Preparation of partially saturated thiazolo- and thiazino[2,3-b]quinazolinone derivatives, and asymmetric 2-[2′-(ethoxycarbonyl-cycloalkyl and -cycloalkenyl)-imino]-3-(2′′-thiazolin-2′′-yl)-thiazolidine diadducts.
  • Synthesis of 3-(β-D-ribofuranosyl)-6,7-dihydrothiazolo[3,2-a]purin-9-one.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zbigniew Fijałek et al.
Journal of pharmaceutical and biomedical analysis, 32(4-5), 967-974 (2003-08-06)
Cyclic voltammetry (CV) and electrochemical quartz crystal microbalance (EQCM) methods were used to study the behavior of sulforaphane (SFR). To established electrochemical properties of SFR methyl-, ethyl-, 2-chloroethyl-, propyl-, butyl- and tert-butyl-isothiocyanates were examined at gold electrode. Optimal measurement parameters
Synthesis and structure determination of alicycle-fused thiazolo-and thiazino [2, 3-bquinazolones and cycloalkylimino-3 (N)-thiazolinylthiazolidines.
Huber I, et al.
Tetrahedron, 48(23), 4949-4956 (1992)
Heterocyclic aromatic anions with 4n+ 2p-electrons.
Bordwell FG and Fried HE.
The Journal of Organic Chemistry, 56(13), 4218-4223 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service