Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400661

Sigma-Aldrich

Sodium zirconate

−200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2ZrO3
CAS Number:
Molecular Weight:
185.20
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Pricing and availability is not currently available. Contact Technical Service

form

powder

particle size

−200 mesh

SMILES string

[Na+].[Na+].[O-][Zr]([O-])=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Bastow et al.
Solid state nuclear magnetic resonance, 3(2), 49-57 (1994-04-01)
A combination of X-ray and electron diffraction, electron microscopy and solid-state nuclear magnetic resonance (NMR) has been used to elucidate the structure and the ordering of Na2ZrO3. The diffraction data confirm a monoclinic crystal structure. A sample prepared by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service