Skip to Content
MilliporeSigma

381470

Sigma-Aldrich

Pentaerythritol tetrakis(2-mercaptoacetate)

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HSCH2CO2CH2)4C
CAS Number:
Molecular Weight:
432.55
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Pricing and availability is not currently available.

grade

technical grade

refractive index

n20/D 1.547 (lit.)

bp

250 °C/1 mmHg (lit.)

density

1.385 g/mL at 25 °C (lit.)

SMILES string

SCC(=O)OCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS

InChI

1S/C13H20O8S4/c14-9(1-22)18-5-13(6-19-10(15)2-23,7-20-11(16)3-24)8-21-12(17)4-25/h22-25H,1-8H2

InChI key

RUDUCNPHDIMQCY-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhong et al.
ACS applied materials & interfaces, 11(32), 28968-28977 (2019-07-25)
A slightly cross-linked lithium borate containing single ion-conducting polymer (LBSIP) as a bifunctional binder for lithium sulfur batteries is designed and fabricated via a one-step thiol-ene click reaction. The LBSIP binder exhibits a maximum peeling strength of over 600 mN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service