Skip to Content
Merck
All Photos(1)

Key Documents

328995

Sigma-Aldrich

2-(4-Hydroxyphenoxy)propionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC6H4OH)CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
2263788
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

143-146 °C (lit.)

SMILES string

CC(Oc1ccc(O)cc1)C(O)=O

InChI

1S/C9H10O4/c1-6(9(11)12)13-8-4-2-7(10)3-5-8/h2-6,10H,1H3,(H,11,12)

InChI key

AQIHDXGKQHFBNW-UHFFFAOYSA-N

General description

2-(4-Hydroxyphenoxy)propionic acid is an intermediate in the synthesis of enantiomerically pure aryloxyphenoxypropionic acid-type herbicides.[1] Enzymatic resolution of (R,S)-2-(4-hydroxyphenoxy)propionic acid was reported.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective preparation of (< i> R</i>)-2-(4-hydroxyphenoxy) propionic acid with a fungal peroxygenase.
Kinne M, et al.
Tetrahedron Letters, 49(41), 5950-5953 (2008)
Enzymatic resolution of (R, S)-2-(4-hydroxyphenoxy) propionic acid.
Barton MJ, et al.
Enzyme and Microbial Technology, 12(8), 577-583 (1990)
Ye Yuan et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 50(8), 533-543 (2015-06-13)
The metabolic fate of (14)C-phenyl-labeled herbicide clodinafop-propargyl ((14)C-CfP) was studied for 28 days in lab assays using a sediment-water system derived from a German location. Mineralization was 5.21% of applied (14)C after 28 days exhibiting a distinct lag phase until

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service