Skip to Content
Merck
All Photos(1)

Documents

288152

Sigma-Aldrich

Sodium hexachloroplatinate(IV) hexahydrate

98%

Synonym(s):

Platinum(IV) sodium chloride, Sodium platinic chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2PtCl6 · 6H2O
CAS Number:
Molecular Weight:
561.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

crystalline

SMILES string

O.O.O.O.O.O.[Na+].[Na+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2Na.6H2O.Pt/h6*1H;;;6*1H2;/q;;;;;;2*+1;;;;;;;+4/p-6

InChI key

ZBFQOIBWJITQRI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

The product may be used as a precursor , for the photoreductive deposition of metallic platinum on various substrates such as a photoactive protein film. These films have potential applications as biohybrid catalysts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodeposition and characterization of thin-film platinum-iridium alloys for biological interfaces
Petrossians A, et al.
Journal of the Electrochemical Society, 158.5, D269-D276 (2011)
Ruhui Chen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(8), 2760-2766 (2020-11-06)
Noble-metal nanocrystals with anisotropic shapes have received increasing interest owing to their unique properties. Here, a facile route to the preparation of Pt nanobars with aspect ratios tunable up to 2.1 was reported by simply reducing a PtIV precursor in
Photoreduction of catalytic platinum particles using immobilized multilayers of photosystem I.
LeBlanc G, et al.
Langmuir, 28.21, 7952-7956 (2012)
Zhuangqiang Gao et al.
ACS nano, 14(1), 791-801 (2020-01-10)
The ability to produce a diverse spectrum of hollow nanostructures is central to the advances in many current and emerging areas of technology. Herein, we report a general method to craft hollow nanostructures with highly tunable physical and chemical parameters.
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service