Skip to Content
Merck
All Photos(1)

Documents

237469

Sigma-Aldrich

Poly(4-vinylpyridinium dichromate)

cross-linked

Synonym(s):

PVPDC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

crosslinking

cross-linked

Application

Convenient polymeric oxidizing agent. The polymer is activated prior to use by soaking in water and filtering. Recycling is achieved by washing the reagent with acid.

Other Notes

To activate, soak in water and filter before using!

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 46, 1728-1728 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service