Skip to Content
Merck
All Photos(2)

Documents

227404

Sigma-Aldrich

Chirald®

99%

Synonym(s):

(2S,3R)-(+)-4-Dimethylamino-3-methyl-1,2-diphenyl-2-butanol, (2S,3R)-(+)-4-Dimethylamino-1,2-diphenyl-3-methyl-2-butanol, Dextrocarbinol base

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C(C6H5)(OH)CH(CH3)CH2N(CH3)2
CAS Number:
Molecular Weight:
283.41
Beilstein:
2384178
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

99%

form

solid

optical activity

[α]21/D +8.2°, c = 10 in ethanol

mp

54-56 °C (lit.)

SMILES string

C[C@H](CN(C)C)[C@@](O)(Cc1ccccc1)c2ccccc2

InChI

1S/C19H25NO/c1-16(15-20(2)3)19(21,18-12-8-5-9-13-18)14-17-10-6-4-7-11-17/h4-13,16,21H,14-15H2,1-3H3/t16-,19+/m1/s1

InChI key

INTCGJHAECYOBW-APWZRJJASA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Chirald is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service