Skip to Content
Merck
All Photos(1)

Documents

190721

Sigma-Aldrich

4-Ethoxy-2-nitroaniline

97%

Synonym(s):

2-Nitro-p-phenetidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(NO2)NH2
CAS Number:
Molecular Weight:
182.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

111-113 °C (lit.)

SMILES string

CCOc1ccc(N)c(c1)[N+]([O-])=O

InChI

1S/C8H10N2O3/c1-2-13-6-3-4-7(9)8(5-6)10(11)12/h3-5H,2,9H2,1H3

InChI key

ISFYBUAVOZFROB-UHFFFAOYSA-N

Application

4-Ethoxy-2-nitroaniline may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Bilic
Journal of chromatography, 543(2), 367-374 (1991-05-10)
A procedure for the simultaneous determination of both ascorbic and dehydroascorbic acid in dairy foods by high-performance liquid chromatography using precolumn derivatization with 4-methoxy- and 4-ethoxy-1,2-phenylenediamine is presented. The derivatives are isolated by solid-phase extraction and analysed by fluorescence detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service