Skip to Content
Merck
All Photos(1)

Key Documents

111945

Sigma-Aldrich

3,4-Dichlorobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3COCl
CAS Number:
Molecular Weight:
209.46
Beilstein:
607485
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050528
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

242 °C (lit.)

mp

30-33 °C (lit.)

SMILES string

ClC(=O)c1ccc(Cl)c(Cl)c1

InChI

1S/C7H3Cl3O/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H

InChI key

VTXNOVCTHUBABW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dichlorobenzoyl chloride was used in the synthesis of 3,4-dichlorophenylacetic acid by Arndt-Eistert method. It is prepared by refluxing 3,4-dichlorobenzoic acid with thionyl chloride.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitors of the Hill reaction.
N E Good
Plant physiology, 36(6), 788-803 (1961-11-01)
Hydroxyindole-O-methyltransferase. 3. Influ- ence of the phenyl moiety on the inhibitory activities of some N-acyltryptamines.
B T Ho et al.
Journal of pharmaceutical sciences, 58(5), 563-566 (1969-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service