Skip to Content
Merck
All Photos(1)

Key Documents

AC7326370154

Advanced Chemical Intermediates Ltd.

3-(2-Chloro-6-fluoro-phenyl)-5-methyl-isoxazole-4-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
269.66
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

methyl 3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate

Inchi Code

1S/C12H9ClFNO3/c1-6-9(12(16)17-2)11(15-18-6)10-7(13)4-3-5-8(10)14/h3-5H,1-2H3

InChI key

VUXXAVIGDIIYBQ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

White crystalline solid

Product Link

https://www.acints.com/products/SB-05F-001

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.acints.com/products/SB-05F-001


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service