Skip to Content
Merck
All Photos(1)

Key Documents

AC7300012896

Advanced Chemical Intermediates Ltd.

5-Bromo-3-methylbenzo[b]thiophene-2-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
325.63
MDL number:

Iupac Name

5-bromo-3-methyl-1-benzothiophene-2-sulfonyl chloride

Inchi Code

1S/C9H6BrClO2S2/c1-5-7-4-6(10)2-3-8(7)14-9(5)15(11,12)13/h2-4H,1H3

InChI key

YSXQHKIUGBLJKG-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Beige solid

Product Link

https://www.acints.com/products/ACI-NI-00004

Pictograms

Signal Word

Danger

Hazard Statements

MSDS

https://www.acints.com/products/ACI-NI-00004


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service