Skip to Content
Merck
All Photos(1)

Key Documents

AC7300012751

Advanced Chemical Intermediates Ltd.

2-(2,6-Dichloro-benzylsulfanyl)-ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
236.16
MDL number:

Iupac Name

2-[(2,6-dichlorobenzyl)sulfanyl]ethanamine

Inchi Code

1S/C9H11Cl2NS/c10-8-2-1-3-9(11)7(8)6-13-5-4-12/h1-3H,4-6,12H2

InChI key

VCKWYLGEZYGACK-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Pale yellow low melting solid

Product Link

https://www.acints.com/products/ACI-00700

Pictograms

Signal Word

Danger

Hazard Statements

MSDS

https://www.acints.com/products/ACI-00700


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service