Skip to Content
Merck
All Photos(1)

Key Documents

AABH99A4DB07

AA BLOCKS, INC.

Octanoic acid, 8-[[(1,1-dimethylethoxy)carbonyl]amino]-, 2,5-dioxo-1-pyrrolidinyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
356.42
MDL number:

Pricing and availability is not currently available.

Iupac Name

2,5-dioxopyrrolidin-1-yl 8-((tert-butoxycarbonyl)amino)octanoate

Inchi Code

1S/C17H28N2O6/c1-17(2,3)24-16(23)18-12-8-6-4-5-7-9-15(22)25-19-13(20)10-11-14(19)21/h4-12H2,1-3H3,(H,18,23)

InChI key

VWJIIRILMYJMEF-UHFFFAOYSA-N

storage temp.

2~8C

Purity

95%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/149414-80-2

Synonym

Octanoic acid, 8-[[(1,1-dimethylethoxy)carbonyl]amino]-, 2,5-dioxo-1-pyrrolidinyl ester

MSDS

https://www.aablocks.com/downloadSds?cas=149414-80-2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service