Iupac Name
(2S)-2-[({(2S)-1-[(2S)-2-({(2S)-4-amino-2-[({(2S)-1-[(2S)-2-amino-3-methylbutanoyl]-2-pyrrolidinyl}carbonyl)amino]-4-oxobutanoyl}amino)-4-methylpentanoyl]-2-pyrrolidinyl}carbonyl)amino]-N~1~-((1S)-1-(
Inchi Code
1S/C45H72N14O10/c1-24(2)21-31(57-40(65)30(23-35(47)61)56-42(67)33-15-10-20-59(33)44(69)36(48)25(3)4)43(68)58-19-9-14-32(58)41(66)54-28(16-17-34(46)60)39(64)53-27(13-8-18-52-45(50)51)38(63)55-29(37(49)62)22-26-11-6-5-7-12-26/h5-7,11-12,24-25,27-33,36H,8-10,13-23,48H2,1-4H3,(H2,46,60)(H2,47,61)(H2,49,62)(H,53,64)(H,54,66)(H,55,63)(H,56,67)(H,57,65)(H4,50,51,52)/t27-,28-,29-,30-,31-,32-,33-,36-/m0/s1
InChI key
JFZCLMZUABKABL-GVCDGELBSA-N
storage temp.
Purity
Country of Origin
Product Link
Synonym
MSDS
https://www.aablocks.com/downloadSds?cas=311309-27-0
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service