Skip to Content
Merck
All Photos(1)

Key Documents

AABH97D2A902

AA BLOCKS, INC.

L-Glutamic acid, L-phenylalanyl-L-isoleucyl-L-α-aspartyl-L-prolyl-L-α-glutamyl-L-leucyl-L-glutaminyl-L-arginyl-L-seryl-L-tryptophyl-L-α-glutamyl-L-α-glutamyl-L-lysyl-L-α-glutamylglycyl-L-α-glutamylglycyl-L-valyl-L-leucyl-L-methionyl-L-prolyl-

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CA$56.30
100 ML
CA$60.20
500 ML
CA$143.00
4 X 100 ML
CA$200.00
1 L
CA$235.00
2.5 L
CA$437.00

CA$56.30


Please contact Customer Service for Availability


Select a Size

Change View
5 ML
CA$56.30
100 ML
CA$60.20
500 ML
CA$143.00
4 X 100 ML
CA$200.00
1 L
CA$235.00
2.5 L
CA$437.00

About This Item

CAS Number:
Molecular Weight:
2618.9
MDL number:

CA$56.30


Please contact Customer Service for Availability

Iupac Name

(4S,10S,13S,16S,19S,22S,25S,28S,31S,34S,37S)-22-((1H-indol-3-yl)methyl)-37-((((S)-1-((S)-2-(((2S,3S)-2-(((S)-2-amino-1-hydroxy-3-phenylpropylidene)amino)-1-hydroxy-3-methylpentylidene)amino)-3-carboxypropanoyl)pyrrolidin-2-yl)(hydroxy)methylene)amino)-13-(4-aminobutyl)-10,16,19-tris(2-carboxyethyl)-4-((6S,9S,12S)-12-((S)-2-((((S)-1,3-dicarboxypropyl)imino)(hydroxy)methyl)pyrrolidine-1-carbonyl)-1,4,7,10-tetrahydroxy-9-isobutyl-6-isopropyl-15-thia-2,5,8,11-tetraazahexadeca-1,4,7,10-tetraen-1-yl)-28-(3-guanidinopropyl)-6,9,12,15,18,21,24,27,30,33,36-undecahydroxy-31-(3-hydroxy-3-iminopropyl)-25-(hydroxymethyl)-34-isobutyl-5,8,11,14,17,20,23,26,29,32,35-undecaazatetraconta-5,8,11,14,17,20,23,26,29,32,35-undecaenedioic acid

Inchi Code

1S/C116H176N28O39S/c1-10-61(8)95(142-96(163)65(118)51-62-21-12-11-13-22-62)112(179)139-80(53-93(161)162)114(181)144-47-20-27-82(144)109(176)133-74(34-41-91(157)158)104(171)136-77(49-58(2)3)105(172)131-71(29-36-84(119)146)101(168)128-68(26-18-45-122-116(120)121)100(167)140-81(57-145)108(175)137-79(52-63-54-123-66-24-15-14-23-64(63)66)107(174)132-73(33-40-90(155)156)103(170)130-72(32-39-89(153)154)102(169)127-67(25-16-17-44-117)99(166)129-70(31-38-88(151)152)98(165)124-55-85(147)126-69(30-37-87(149)150)97(164)125-56-86(148)141-94(60(6)7)111(178)138-78(50-59(4)5)106(173)134-75(43-48-184-9)113(180)143-46-19-28-83(143)110(177)135-76(115(182)183)35-42-92(159)160/h11-15,21-24,54,58-61,65,67-83,94-95,123,145H,10,16-20,25-53,55-57,117-118H2,1-9H3,(H2,119,146)(H,124,165)(H,125,164)(H,126,147)(H,127,169)(H,128,168)(H,129,166)(H,130,170)(H,131,172)(H,132,174)(H,133,176)(H,134,173)(H,135,177)(H,136,171)(H,137,175)(H,138,178)(H,139,179)(H,140,167)(H,141,148)(H,142,163)(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H,159,160)(H,161,162)(H,182,183)(H4,120,121,122)/t61-,65-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,94-,95-/m0/s1

InChI key

ICAYDYVQHKWGMZ-VIAQXQIWSA-N

storage temp.

2~8C

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/122018-92-2

Synonym

CORTICOTROPIN RELEASE-INHIBITING FACTOR

MSDS

https://www.aablocks.com/downloadSds?cas=122018-92-2


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service