Key Documents
Z722390
Sigma-Aldrich
x-tracta Gel Extraction Tool
x-tracta Gel Extraction Tool
1 of 1 reviewers received a sample product or took part in a promotion
pkg of 100 ea
Synonym(s):
dna extraction, dna extraction tool, dna isolation, dna isolation tool, nucleic acid isolation, rna isolation, rna isolation tool
Select a Size
100 EA
$135.00
$135.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EA
$135.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
$135.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 100 ea
General description
Benefits of the x-tracta Gel Extraction Tool:
  • Fast
  • East to Use
  • One-Handed Procedure
  • Precise Extraction
  • High Yield Recovery
  • Disposable
  • Safe
For an instructional overview and further details regarding the x-tracta gel extraction tool, please visit www.sigmaaldrich.com/xtracta
The x-tracta gel extraction tool is an easy to use, quick alternative to using scalpel or razor blades for the extraction of DNA and RNA gel bands from agarose gels following gel electrophoresis. The cutting blade and ergonomic design mean that nucleic acid agarose gel extractions can be performed in a rapid, easy to follow process with greater accuracy and precision. Simply stated, the superior cutting edge of x-tracta along with its easy, one-handed operation means that it performs better than any nucleic acid gel extraction solution for agarose gel band extraction that is currently available.
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
A quick, clean and easy to use alternative to using a scalpel or razor blade for the extraction of DNA and RNA bands from agarose gels following gel electrophoresis.
The GenElute Gel Extraction kit is designed for the rapid purification of 50 bp to 10 kb linear DNA fragments and plasmids from standard or low-melting agarose gels.
Questions
Reviews
1 of 1 reviewers received a sample product or took part in a promotion
Active Filters
  1. California
    • Review 1
    • Votes 2
    5 out of 5 stars.
    Works extremely well
    These devices are really slick for removing gel slices for sequencing. Their only drawback is their cost.
    Helpful?