Skip to Content
Merck
All Photos(1)

Key Documents

Z361569

Extractor for Ethidium Bromide Decontamination

includes storage cap and adhesive label

Synonym(s):

Ethidium Bromide Extractor

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$226.00
6 EA
$632.00

$226.00


Estimated to ship onMay 04, 2025



Select a Size

Change View
2 EA
$226.00
6 EA
$632.00

About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

$226.00


Estimated to ship onMay 04, 2025


manufacturer/tradename

Maine Mfg 10448030

Application

The extractor for ethidium bromide decontamination is a self-contained activated carbon extraction device for removing mutagenic ethidium bromide (EtBr) from electrophoresis buffers. Each unit processes a liter of buffer in less than a minute, removing >99% of EtBr and maintains this efficiency for up to 10 liters [initial (EtBr) approx. 0.5 mg/liter]. Assuming no other environmentally hazardous components, processed buffer can be discarded down the laboratory drain.
Fits most standard laboratory flasks. Includes a cap for storage between uses and an adhesive label to log usage 0.5 liter at a time.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service