Skip to Content
Merck
All Photos(2)

Key Documents

T0303

Sigma-Aldrich

Trypsin from porcine pancreas

Trypsin from porcine pancreas

Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein

Synonym(s):

Serine Protease 1

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$230.00
10 G
$1,130.00

$230.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$230.00
10 G
$1,130.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.78

$230.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

type

Type IX-S

Quality Level

form

lyophilized powder

specific activity

13,000-20,000 BAEE units/mg protein

mol wt

23.8 kDa

application(s)

diagnostic assay manufacturing

foreign activity

chymotrypsin ≤1.0 units/mg protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestionsns†. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Components

Trypsin consists of a single chain polypeptide of 223 amino acid residues, produced by the removal of the N-terminal hexapeptide from trypsinogen which is cleaved at the Lys - lle peptide bond. The sequence of amino acids is cross-linked by 6 disulfide bridges. This is the native form of trypsin, beta-trypsin. BETA-trypsin can be autolyzed, cleaving at the Lys - Ser residue, to produce alpha-trypsin. Trypsin is a member of the serine protease family.

Caution

Solutions in 1 mM HCl are stable for 1 year in aliquots and stored at -20°C. The presence of Ca2+ will also diminish the self-autolysis of trypsin and maintain its stability in solution. Trypsin will also retain most of its activity in 2.0 M urea, 2.0 M guanidine HCl, or 0.1% (w/v) SDS.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25° C using BAEE as substrate. One BTEE unit = 320 ATEE units. Reaction volume = 3.2 mL (1 cm light path).

Preparation Note

This product is a lyophilized powder soluble in Hank′s Balanced Salt Solution at 25 mg/mL.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelle Matthijnssens et al.
Journal of virology, 84(4), 2013-2026 (2009-11-27)
Although few simian rotaviruses (RVs) have been isolated, such strains have been important for basic research and vaccine development. To explore the origins of simian RVs, the complete genome sequences of strains PTRV (G8P[1]), RRV (G3P[3]), and TUCH (G3P[24]) were
S Rajeshkumar et al.
Fish & shellfish immunology, 26(3), 429-437 (2009-01-15)
The protective efficacy of oral delivery of a DNA construct containing the VP28 gene of WSSV encapsulated in chitosan nanoparticles was investigated in black tiger shrimp (Penaeus monodon). The results showed that significant survival was obtained in WSSV-challenged shrimp at
G K German et al.
Biophysical journal, 102(11), 2424-2432 (2012-06-21)
We study the drying of stratum corneum, the skin's outermost layer and an essential barrier to mechanical and chemical stresses from the environment. Even though stratum corneum exhibits structural features across multiple length-scales, contemporary understanding of the mechanical properties of
Arno G B Wouters et al.
Journal of agricultural and food chemistry, 65(6), 1263-1271 (2017-01-27)
The relationship between structural and foaming properties of two tryptic and two peptic wheat gluten hydrolysates was studied at different pH conditions. The impact of pH on foam stability (FS) of the samples heavily depended on the peptidase used and
Valeria Lulla et al.
Nature communications, 11(1), 4070-4070 (2020-08-15)
Human astroviruses are small non-enveloped viruses with positive-sense single-stranded RNA genomes. Astroviruses cause acute gastroenteritis in children worldwide and have been associated with encephalitis and meningitis in immunocompromised individuals. It is still unknown how astrovirus particles exit infected cells following

Protocols

Continuous spectrophotometric rate determination method using BAEE substrate measures trypsin activity, essential for enzyme characterization.

Questions

Reviews

Active Filters

  1. Blacksburg VA
    • Review 1
    • Votes 2
    4 out of 5 stars.

    Active but consider autoproteolysis

    Trypsin is active but autoproteolyze over time. It requires re-purification and design to reduce autoproteolysis.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service