Skip to Content
Merck
All Photos(2)

Key Documents

L3042

Sigma-Aldrich

Levocabastine hydrochloride

≥99% (HPLC), solid

Synonym(s):

Levophta, Levostin, R50547, [3S-[1(cis),3α,4β]]-1-[4-cyano-4-(4-fluorophenyl)cyclohexyl]-3-methyl-4-phenyl-4-piperidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29N2O2F·HCl
CAS Number:
Molecular Weight:
456.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (HPLC)

form

solid

color

white

solubility

DMSO: soluble ~10 mg/mL

originator

Johnson & Johnson

SMILES string

Cl[H].C[C@@H]1CN(CC[C@]1(C(O)=O)c2ccccc2)[C@@H]3CC[C@@](CC3)(C#N)c4ccc(F)cc4

InChI

1S/C26H29FN2O2.ClH/c1-19-17-29(16-15-26(19,24(30)31)21-5-3-2-4-6-21)23-11-13-25(18-28,14-12-23)20-7-9-22(27)10-8-20;/h2-10,19,23H,11-17H2,1H3,(H,30,31);1H/t19-,23-,25-,26-;/m1./s1

InChI key

OICFWWJHIMKBCD-VALQNVSPSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-peptide histamine H1 receptor antagonist; neurotensin NTS2 receptor ligand.

Features and Benefits

This compound is featured on the Neurotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Geneviève Roussy et al.
Molecular pain, 5, 38-38 (2009-07-08)
Central neurotensin (NT) administration results in a naloxone-insensitive antinociceptive response in animal models of acute and persistent pain. Both NTS1 and NTS2 receptors were shown to be required for different aspects of NT-induced analgesia. We recently demonstrated that NTS2 receptors
A Leonardi et al.
The British journal of ophthalmology, 91(12), 1662-1666 (2007-06-23)
This comparative and randomised pilot study assessed the clinical and biological efficacy of Naaxia Sine(R) eye-drops versus levocabastine eye-drops in the treatment of vernal keratoconjunctivitis (VKC). Twenty-three VKC patients were randomised and treated bilaterally for 28 days with N-acetyl-aspartyl-glutamate (NAAGA)
Ahmed R Qasem et al.
Biochemical pharmacology, 76(6), 751-762 (2008-08-06)
Levocabastine is an antiallergic drug acting as a histamine H1-receptor antagonist. In allergic conjunctivitis (AC), it may also antagonize up-regulation of the intercellular adhesion molecule-1 (ICAM-1) expressed on epithelial conjunctival cells. However, little is known about its effects on eosinophils
Pascal Tétreault et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(9), 3741-3752 (2013-06-13)
Management of painful peripheral neuropathies remains challenging, since patients with chronic pain respond poorly to the available pharmacopeia. In recent years, the G-protein-coupled receptor neurotensin (NT) type 2 (NTS2) emerged as an attractive target for treating transitory pain states. To
J M Botto et al.
Neuroscience letters, 223(3), 193-196 (1997-02-28)
The effect of the drug SR 48692 on the Ca(2+)-activated Cl- current induced by neurotensin on Xenopus oocytes injected with cRNAs encoding rodent high and low affinity neurotensin receptors, was examined. In this receptor expression system, SR 48692 failed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service