CLS431110
Corning® square bioassay dishes
without handles, TC treated, square
Synonym(s):
cell culture dishes, cell culture plates, culture dish
Select a Size
$200.00
Estimated to ship onApril 12, 2025
Select a Size
About This Item
$200.00
Estimated to ship onApril 12, 2025
Recommended Products
material
polystyrene
square
sterility
sterile; γ-irradiated
packaging
sleeve of 4 x 4
manufacturer/tradename
Corning 431110
H
25 mm
L × W
245 mm × 245 mm
surface area
500 cm2
General description
- Corning® square bioassay dishes are made from optically clear polystyrene.
- Stacking bead design facilitate secure stacking and prevents slipping.
- Compatible with automated colony picking instruments.
- Certified nonpyrogenic dishes are supplied with lids.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)