Skip to Content
Merck
All Photos(4)

Key Documents

C7926

Sigma-Aldrich

Collagenase from Clostridium histolyticum

Sigma Blend Type F, ≥2.0 FALGPA units/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$99.50
100 MG
$216.00
500 MG
$856.00
1 G
$1,340.00
5 G
$8,270.00

$99.50


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$99.50
100 MG
$216.00
500 MG
$856.00
1 G
$1,340.00
5 G
$8,270.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$99.50


Estimated to ship onMay 23, 2025


Request a Bulk Order

biological source

Clostridium histolyticum

Quality Level

form

powder

caseinase activity

≤10 units/mg solid

specific activity

≥2.0 FALGPA units/mg solid

mol wt

68-130 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Collagenase from Clostridium histolyticum has been used in determining the degradation rate of collagen biomaterials.[1] It may also be used to determine the enzymatic degradation and enzymatic resistance by the liberated L-leucine measurements.[2]
Collagenase from Clostridium histolyticum, or Clostridiopeptidase A, has been used in a study to assess contact dermatitis with clostridiopeptidase A contained in Noruxol ointment. Clostridiopeptidase A has also been used in a study to investigate the early and successful enzymatıc debridement via collagenase application to pinna in a preterm neonate.
The enzyme has been used in the isolation of mast cells from adult mice. This study generated a large number of connective tissue-type mast cells by the culture of murine fetal skin cells.[3] The product has also been used in the in vitro biodegradation study of Bombyx mori silk fibroin fibers and films, by exposing them to the enzyme for varied time periods.[4]

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid (EGTA)4; β-mercaptoethanol; glutathione, reduced; thioglycolic acid, sodium; and 2,2′-dipyridyl; 8-hydroxyquinoline are known to inhibit the enzyme activity.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuo Yamada et al.
The Journal of investigative dermatology, 121(6), 1425-1432 (2003-12-17)
We describe a novel culture system for generating large numbers of murine skin-associated mast cells and distinguish their characteristics from bone marrow-derived cultured mast cells. Culture of day 16 fetal skin single cell suspensions in the presence of interleukin-3 and
Biodegradation of Bombyx mori silk fibroin fibers and films.
Arai T, et al.
Journal of Applied Polymer Science, 91(4), 2383-2390 (2004)
Properties of collagen and hyaluronic acid composite materials and their modification by chemical crosslinking
Rehakova M, et al.
Journal of Biomedical Materials Research, 30(3), 369-372 (1996)
K Vizárová et al.
Biomaterials, 16(16), 1217-1221 (1995-11-01)
Two kinds of layered atelocollagen materials cross-linked with hexamethylene diisocyanate (HMDIC), starch dialdehyde and glyoxal were enzymatically treated by bacterial collagenase. Evaluating collagenase digestion assay for these material showed progressive differences, particularly in the group of samples cross-linked with HMDIC.
Brittney R Garner et al.
The Journal of pharmacology and experimental therapeutics, 376(1), 40-50 (2020-10-27)
Pharmacological openers of ATP-sensitive potassium (KATP) channels are effective antihypertensive agents, but off-target effects, including severe peripheral edema, limit their clinical usefulness. It is presumed that the arterial dilation induced by KATP channel openers (KCOs) increases capillary pressure to promote

Protocols

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service