Skip to Content
Merck
All Photos(1)

Key Documents

B2064

Sigma-Aldrich

Bovine Serum Albumin

heat shock fraction, Australia origin, protease free, low fatty acid, low IgG, pH 7, ≥98%

Synonym(s):

Bovine Serum Albumin, Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

bovine

Quality Level

200
300

Assay

≥98%

form

powder

mol wt

~66 kDa

purified by

heat shock fractionation

origin

Australia origin

Fatty acid content

<0.05%

technique(s)

immunofluorescence: suitable

impurities

≤0.05% IgG

pH

7

foreign activity

protease ≤0.05 U/mg

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA) is made up of 583 amino acids, with no carbohydrates.[1][2] It is a water-soluble protein with a molecular weight of 66.4kDa. Six α-helices form three homologous domains of BSA. Depending on pH, it undergoes reversible conformational isomerization. The native structure of BSA becomes reactive and flexible on heating.[3] BSA acts as a model protein in various studies, such as immunodiagnostic procedures, cell culture media and clinical chemistry. It is also relevant in foods containing bovine milk or meat.[2]
Appl- Bovine serum albumin has been used as a supplement of phosphate buffered saline (PBS) to store brain tissue sections.[4] It has also been used as a blocking agent for cells for immunofluorescence analysis.[5][6]

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Australia origin
  • Fatty acid free / ultra-low fatty acid content (≤ 0.05%)
  • Protease-free
  • Low IgG content (≤ 0.05% IgG by Ouchterlony Titer)

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia induces pulmonary arterial fibroblast proliferation, migration, differentiation and vascular remodeling via the PI3K/Akt/p70S6K signaling pathway.
Chai X, et al.
International Journal of Molecular Medicine, 41(5), 2461-2472 (2018)
Effect of Linguizhugan decoction on neuroinflammation and expression disorder of the amyloid β-related transporters RAGE and LRP-1 in a rat model of Alzheimer's disease.
Hu Qianfeng, et al.
Molecular Medicine Reports, 17(1), 827-834 (2018)
Kinetics of Thermal Denaturation and Aggregation of Bovine Serum Albumin.
Borzova VA, et.al.
PLoS ONE, 11(4), e0153495-e0153495 (2016)
Jiawei Liu et al.
Analytical and bioanalytical chemistry, 410(2), 573-584 (2017-12-01)
Polymeric ionic liquids (PILs) with 1-vinyl-3-ethylimidazolium cations and two different anions of Br
Rapid confirmation and revision of the primary structure of bovine serum albumin by ESIMS and Frit-FAB LC/MS.
Hirayama K, et.al.
Biochemical and Biophysical Research Communications, 173(2), 639-646 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service