Skip to Content
Merck
All Photos(2)

Key Documents

A8582

Sigma-Aldrich

Antifoam SE-15

aqueous emulsion for bacterial and mammalian systems

Synonym(s):

silicone-based antifoam

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161901
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

form

emulsion (aqueous)

concentration

10% (active silicon)

technique(s)

cell culture | hybridoma: suitable

solubility

H2O: soluble (effective hot or cold.)

density

1.0 g/mL at 25 °C

application(s)

microbiology

storage temp.

room temp

suitability

(Mammalian (suspension))
bacteria (fermentation)

General description

Antifoam SE-15 is a silicone-based product used to prevent or minimize foaming in both microbial fermentation systems and mammalian suspension cultures. Control of foaming minimizes damage to the cells, resulting in increased protein/antibody production.

Application

Antifoam SE-15 has been used:

  • in the preparation of ramified rolling circle amplification (RAM) amplification buffer.[1]
  • to find its effects on cell viability, virus infectivity and PCR (polymerase chain reaction) performance.[2]
  • as a constituent of phosphate-buffered saline (PBS) for the determination of total antioxidant status (TAOS) by 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid (ABTS) assay. [3]
  • as a component of autoinduction expression culture. [4]
  • as a component of cyclic adenosine monophosphate (cAMP) detection buffer. [5]

Features and Benefits

  • Controls foaming in microbial culture flasks or fermentations
  • Non-toxic to the culture medium
  • Can be pumped to fermenter on an as-needed basis

Components

Antifoam SE-15 is an aqueous emulsion containing 10% active silicon and non-ionic emulsifiers .

Preparation Note

Antifoam SE-15 can be pre-diluted with 3-10 parts of cool water to aid in dispersion. Pre-diluted
suspensions should be used immediately.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Lungs

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Detection of nucleic acid targets using ramified rolling circle DNA amplification: a single nucleotide polymorphism assay model
Smith J H and Beals T P
PLoS ONE, 8(5), e65053-e65053 (2013)
Elisa Binda et al.
BMC biotechnology, 13, 24-24 (2013-03-19)
VanYn, encoded by the dbv7 gene (also known as vanYn) of the biosynthetic cluster devoted to A40926 production, is a novel protein involved in the mechanism of self-resistance in Nonomuraea sp. ATCC 39727. This filamentous actinomycete is an uncommon microorganism
James H Smith et al.
PloS one, 8(5), e65053-e65053 (2013-06-01)
Isothermal amplification methods provide alternatives to PCR that may be preferable for some nucleic acid target detection tasks. Among current isothermal target detection methods, ramified rolling circle amplification (RAM) of single-stranded DNA circles that are formed by ligation of linear
Anna Katharina Schuh et al.
Redox report : communications in free radical research, 23(1), 83-93 (2017-12-20)
Redox regulation plays a crucial role in balancing the cardiovascular system. In this prospective study we aimed to identify currently unknown correlations valuable to cardiovascular research and patient management. Blood samples from 500 patients were collected directly before cardiosurgical interventions
Dario A Vitturi et al.
American journal of physiology. Heart and circulatory physiology, 296(5), H1398-H1407 (2009-03-17)
Allosteric regulation of nitrite reduction by deoxyhemoglobin has been proposed to mediate nitric oxide (NO) formation during hypoxia. Nitrite is predominantly an anion at physiological pH, raising questions about the mechanism by which it enters the red blood cell (RBC)

Questions

1–3 of 3 Questions  
  1. 細胞培養液に添加するためにアンチフォームSE-15の購入を検討しております。この商品は、オートクレーブ可能でしょうか、オートクレーブ可能の場合何回程度が限度でしょうか。 もし可能であれば、アンチフォーム C エマルジョンとの違いも教えていただけないでしょうか。 (アンチフォーム C エマルジョンもオートクレーブ可能でしょうか。) よろしくお願い申し上げます。

    1 answer
    1. Antifoam SE-15 is a 10% emulsion of active silicone polymer and non-ionic emulsifiers. It can be repeatedly sterilized by autoclaving. Antifoam C Emulsion is an aqueous emulsion containing 30% active silicone. Further details about the different properties can be found in the datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/316/313/a8582pis.pdf

      Helpful?

  2. At what concentration is SE-15 (A8582) to be used in bacterial cell culture to control foam?

    1 answer
    1. This Antifoam product is typically used at starting concentrations of 0.01 - 0.02%. Optimization may be required based on variable culture density, ingredients, and conditions. Please see the link below to review the product datasheet for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/133/175/a8582dat.pdf

      Helpful?

  3. I was wondering if you had any information in regards to the stability of antifoam SE-15, A8582. In particular, do you have any data about the stability of this product at pH 8.0? When in a solution, do you have any data on the product stability?

    1 answer
    1. This product is stable at room temperature. Antifoam SE-15 can be pre-diluted with 3-10 parts of cool water to aid in dispersion. Pre-diluted suspensions should be used immediately. The stability of this product is not tested at pH 8.0. If you are looking for antifoams that will be stable at pH 8.0, perhaps you may consider Antifoam A Concentrate (Product A5633 and A6582), They are stable at pH 5.0-9.0. Please see this link for any additional information on Product A8582: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/316/313/a8582pis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service