Skip to Content
Merck
All Photos(1)

Key Documents

A6457

Sigma-Aldrich

Antifoam Y-30 Emulsion

aqueous emulsion for bacterial and mammalian systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

description

Molecular Biology Reagent

form

emulsion (aqueous)

concentration

30% (active silicon)

technique(s)

cell culture | hybridoma: suitable

density

1.0 g/mL at 25 °C

application(s)

microbiology

storage temp.

room temp

suitability

(Mammalian (suspension))
bacteria (fermentation)

General description

Antifoam Y-30 is a silicone-based product used to prevent or minimize foaming in both microbial fermentation systems and mammalian suspension cultures. Control of foaming minimizes damage to the cells, resulting in increased protein/antibody production.

Application

Suitable for preventing or minimizing foaming in microbial fermentation systems and mammalian suspension cultures.

Components

Antifoam Y-30 is an aqueous emulsion containing 30% active silicon and non-ionic emulsifiers .

Preparation Note

Antifoam Y-30 can be pre-diluted with 3-10 parts of cool water to aid in dispersion. Pre-diluted suspensions should be used immediately.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bonnie Mull et al.
Applied and environmental microbiology, 75(11), 3593-3597 (2009-04-14)
Enterohemorrhagic Escherichia coli O157:H7 (EHEC O157:H7) outbreaks have revealed the need for improved analytical techniques for environmental samples. Ultrafiltration (UF) is increasingly recognized as an effective procedure for concentrating and recovering microbes from large volumes of water and treated wastewater.
Vincent R Hill et al.
Applied and environmental microbiology, 73(13), 4218-4225 (2007-05-08)
Ultrafiltration (UF) is increasingly being recognized as a potentially effective procedure for concentrating and recovering microbes from large volumes of water and treated wastewater. Because of their very small pore sizes, UF membranes are capable of simultaneously concentrating viruses, bacteria
Sigma, BioFiles For Life Science Research: Detergents and Solubilization Reagenter
Sigma data, 3, 33-33 (2008)
Crissy L Tarver et al.
Crystals, 11(10) (2021-11-09)
Among its attributes, the mythical philosopher's stone is supposedly capable of turning base metals to gold or silver. In an analogous fashion, we are finding that protein crystallization optimization using ionic liquids (ILs) often results in the conversion of base
Carmela M Smith et al.
Applied and environmental microbiology, 75(16), 5284-5289 (2009-06-30)
Dead-end ultrafiltration (DEUF) is an alternative approach to tangential-flow hollow-fiber ultrafiltration that can be readily employed under field conditions to recover microbes from water. The hydraulics of DEUF and microbe recovery for a new DEUF method were investigated using 100-liter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service