Skip to Content
Merck
All Photos(1)

Key Documents

A5758

Sigma-Aldrich

Antifoam Y-30 Emulsion

aqueous-silicone emulsion

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900

Quality Level

form

emulsion

contains

emulsifier

General description

Antifoam Y-30 Emulsion is an aqueous emulsion containing 30% active silicon. It contains non-ionic emulsifiers different from those in Antifoam Emulsions B and C. It is designed to control foam in aqueous systems.
Antifoam is also known as defoamer. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. It is used to prevent formation of foam or is added to break foam that is already formed.

Application

Antifoam Y-30 Emulsion is used for the following applications:
  • Chemostat cultivation.[1]
  • Cultivation conditions, such as Fed-batch fermentation.[2][3]
  • Bacterial strains and growth conditions (used in all aerosol experiments).[4]

Other Notes

A 30% aqueous emulsion of Antifoam A concentrate.

Linkage

More easily dispersed in aqueous systems than Antifoam A concentrate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modeling of growth and energy metabolism of Pichia pastoris producing a fusion protein.
M. Jahic, J.C. Rotticci-Mulder, M. Martinelle, K. Hult, S-O. Enfors
Bioprocess and Biosystems Engineering, 24, 385-393 (2002)
Adrien Pagnier et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(1), 104-109 (2015-12-25)
The synthesis and assembly of the active site [FeFe] unit of [FeFe]-hydrogenases require at least three maturases. The radical S-adenosyl-l-methionine HydG, the best characterized of these proteins, is responsible for the synthesis of the hydrogenase CO and CN(-) ligands from
Sarah J Routledge et al.
Methods in molecular biology (Clifton, N.J.), 866, 87-97 (2012-03-29)
Pichia pastoris is a widely used host for recombinant protein production. The foaming associated with culturing it on a large scale is commonly prevented by the addition of chemical antifoaming agents or "antifoams." Unexpectedly, the addition of a range of
Mehmedalija Jahic et al.
Microbial cell factories, 2(1), 6-6 (2003-07-23)
BACKGROUND: A temperature limited fed-batch (TLFB) technique is described and used for Pichia pastoris Mut+ strain cultures and compared with the traditional methanol limited fed-batch (MLFB) technique. A recombinant fusion protein composed of a cellulose-binding module (CBM) from Neocallimastix patriciarum
N Meinander et al.
Microbiology (Reading, England), 142 ( Pt 1), 165-172 (1996-01-01)
Recombinant Saccharomyces cerevisiae harbouring the xylose reductase (XR) gene XYL1 from Pichia stipitis was grown in anoxic chemostat culture at two different dilution rates. At each dilution rate a transient experiment, encompassing a shift in the sugar content of the

Questions

1–7 of 7 Questions  
  1. Are Antifoam A (product number 10794) and Antifoam Y-30 (product number A5758) the same product? Can I use Antifoam Y-30 (product number A5758) as a replacement for Antifoam A (product number 10794)?

    1 answer
    1. The A5758, Antifoam Y-30, is a 30% solution containing non-ionic emulsifiers. It is prepared from an Antifoam A product that does not contain emulsifiers. The 10794, Antifoam A, is a deleted product, but was also a 30% solution containing emulsifiers. These products are from different brands, but functionally are identical and may be used interchangeably.

      Please view the link below for the product information sheet for the offered Antifoam products:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/136/342/a5758pis.pdf

      Helpful?

  2. What is the difference between product A5758, Antifoam Y-30 emulsion and product A6457?

    1 answer
    1. Product A6457, a molecular biology reagent, has been function-tested by Sigma for use in bacterial fermentation. Product A5758 has not been tested in this application by Sigma.

      Helpful?

  3. What is Antifoam Y-30 emulsion?

    1 answer
    1. Antifoam Y-30 emulsion,a Dow-Corning product, is a 30% active silicone emulsion, whose usage is intended for controlling foam in aqueous systems. The composition of the antifoam is considered proprietary by the manufacturer.  However, the emulsifier type is described as nonionic.

      Helpful?

  4. Has there been a name change for Antifoam Y-30 Emulsion, Product A5758?

    1 answer
    1. Yes. The product was formerly listed under the name "Antifoam A Emulsion".  Although there has been a name change, there is no change in the composition of the antifoam. It is the same product.

      Helpful?

  5. Can Antifoam Y-30 Emulsion, Product A5758, be autoclaved?

    1 answer
    1. Dow Corning® does not recommend that the emulsion be autoclaved since autoclaving can adversely affect the product.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. How can Product No. A5758, Antifoam Y-30 Emulsion be used?

    1 answer
    1. This antifoam can be diluted with water. It is effective at low concentrations (1-100 ppm is the usual range to use). Trials can start at 50 ppm (active silicone), and amounts may be reduced to where the level of foam control is desired. The product should be agitated before use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service