A3156
Bovine Serum Albumin
lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Synonym(s):
Albumin bovine serum, BSA, Bovine albumin
About This Item
Recommended Products
biological source
bovine
Quality Level
sterility
γ-irradiated
product line
BioXtra
Assay
≥99% (agarose gel electrophoresis)
form
lyophilized powder
mol wt
~66 kDa
purified by
cold ethanol and heatshock fractionation
origin
USA origin
technique(s)
cell culture | mammalian: suitable
impurities
≤10% water (Karl Fischer)
pH
4.8-7.5
solubility
water: soluble (5g/50ml)
UniProt accession no.
shipped in
ambient
storage temp.
2-8°C
Gene Information
bovine ... ALB(280717)
General description
Application
- to prevent sticking of the chromosome to the glass micropipette for elasticity measurements using aspiration[3]
- to block lung homogenates in the plate for mucin protein enzyme-linked immunosorbent assay[4]
- in alginate solution as a protein model for drug release kinetics investigation[5]
Biochem/physiol Actions
Preparation Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.
Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.
Related Content
Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service