Skip to Content
Merck
All Photos(1)

Key Documents

491969

Sigma-Aldrich

Tris(cyclopentadienyl)yttrium(III)

99.9% trace metals basis

Synonym(s):

YCp3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(C5H5)3
CAS Number:
Molecular Weight:
284.19
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

reaction suitability

core: yttrium
reagent type: catalyst

mp

296 °C (dec.) (lit.)

SMILES string

[Y].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[CH]3[CH][CH][CH][CH]3

InChI

1S/3C5H5.Y/c3*1-2-4-5-3-1;/h3*1-5H;

InChI key

IZLLLZUVMGFBAT-UHFFFAOYSA-N

General description

Atomic number of base material: 39 Yttrium

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the different low temperature direct alcohol fuel cells based on nanostructured material catalysts and anion exchange membrane fuel cells.

Thin film solid oxide fuel cells offer technical advantages; review of TF-SOFC deposition methods for electrolyte thin films.

This article describes the use and advantages of polyoxometalate-based redox-flow batteries as electrochemical energy storage systems over Li-ion batteries.

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service