Skip to Content
Merck
All Photos(1)

Key Documents

324175

Sigma-Aldrich

Lithium 2-thienylcyanocuprate solution

0.25 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3CuLiNS
CAS Number:
Molecular Weight:
179.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

0.25 M in THF

density

0.895 g/mL at 25 °C

SMILES string

[Li]N=C=[Cu]c1cccs1

InChI

1S/C4H3S.CN.Cu.Li/c1-2-4-5-3-1;1-2;;/h1-3H;;;/q;-1;;+1

InChI key

BDZYNOMLDVYLHW-UHFFFAOYSA-N

General description

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.

Application

Useful reagent for the preparation of higher-order cuprates.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aldrichimica Acta, 20, 58-58 (1987)

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service