Skip to Content
Merck
All Photos(1)

Key Documents

254150

Sigma-Aldrich

Bismuth(III) nitrate pentahydrate

99.999% trace metals basis

Synonym(s):

Bismuth trinitrate pentahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$155.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$155.00

About This Item

Linear Formula:
Bi(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
485.07
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.999% trace metals basis

form

crystals

reaction suitability

core: bismuth
reagent type: catalyst

bp

75-80 °C (lit.)

mp

30 °C (lit.)

SMILES string

O=[N+](O[Bi](O[N+]([O-])=O)O[N+]([O-])=O)[O-].C

InChI

1S/CH4.Bi.3NO3/c;;3*2-1(3)4/h1H4;;;;/q;+3;3*-1

InChI key

DPLOIMSBRDLLRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines.[1] Convenient reagent for selective oxidation of sufides to sulfoxides.[2]

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guojun Han et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 91(12), 1150-1163 (2017-12-06)
Mass cytometry (or CyTOF) is an atomic mass spectrometry-based single-cell immunoassay technology, which has provided an increasingly systematic and sophisticated view in basic biological and clinical studies. Using elemental reporters composed of stable heavy metal isotopes, more than 50 cellular
Mashraqui, S.H. Karnik, M.A.
Synthesis, 713-713 (1998)
Mashraqui, S.H. et al.
Synthetic Communications, 28, 939-939 (1998)
Na Li et al.
Dalton transactions (Cambridge, England : 2003), 43(36), 13742-13750 (2014-08-12)
The xFe2O3/yBiOCl composites (xFe/yBi, x/y = 0/100, 5/100, 10/100, 20/100, 30/100 and 40/100 molar ratios) are prepared for the first time through an in situ hydrolysis method under hydrothermal conditions. The samples are characterized by X-ray diffraction (XRD), scanning electron
Nahid Gharib Naseri et al.
Analytical and bioanalytical chemistry, 391(4), 1283-1292 (2008-03-21)
Catalytic adsorptive stripping voltammetry (CAdSV) has been demonstrated at a bismuth film electrode (BiFE) in an injection-moulded electrochemical micro-flow cell. The polystyrene three-electrode flow cell was fabricated with electrodes moulded from a conducting grade of polystyrene containing 40% carbon fibre

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service