Merck
HomeProfessor Product PortalGarg Group – Professor Product Portal
Garg Group – Professor Product Portal
Professor Neil Garg
Professor Neil Garg
The Garg Group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions. Heterocyclic arynes can be trapped by a variety of reagents (e.g., nucleophiles and cycloaddition partners) to prepare diverse libraries of substituted heterocycles from common precursors. The Garg lab has also collaborated with Professor Ken Houk to develop the aryne distortion model, which in turn allows chemists to predict regioselectivity in reactions of unsymmetrical arynes.
Garg Group Website
Recent papers from the Garg Group
1.
Goetz AE, Garg NK. 2013. Regioselective reactions of 3,4-pyridynes enabled by the aryne distortion model. Nature Chem. 5(1):54-60. https://doi.org/10.1038/nchem.1504
2.
Im GJ, Bronner SM, Goetz AE, Paton RS, Cheong PH, Houk KN, Garg NK. 2010. Indolyne Experimental and Computational Studies: Synthetic Applications and Origins of Selectivities of Nucleophilic Additions. J. Am. Chem. Soc.. 132(50):17933-17944. https://doi.org/10.1021/ja1086485
3.
Mesganaw T, Im GJ, Garg NK. 2013. Synthesis of Diphenylhexatriene by the Pd-Catalyzed Dimerization of Cinnamyl Acetate. J. Org. Chem.. 78(7):3391-3393. https://doi.org/10.1021/jo400032j
Sorry, an unexpected error has occurred
Network error: Failed to fetch