Mobius® Essential Assemblies
Ready-To-Ship Single-Use Bag and Tubing Assemblies
SPEED UP YOUR FLUID MANAGEMENT WITH STANDARD DESIGNS AND FAST DELIVERY
Mobius® Essential Assemblies are standard, off-the-shelf bag and tubing assemblies. Made with our proven PureFlex™ film, these plug-and-play assemblies can be easily integrated into your current processes and are available in a variety of configurations, including 1-50 L 2D bag assemblies, 100-200 L 3D bag assemblies, as well as bioprocess tubing assemblies. Products are stocked and will be shipped the next working day!
Key Benefits:
- Eliminate design and manufacturing lead time
- 1 L - 200 L sizes that can easily scale up to premium assemblies
- Flexible, robust, and chemical-resistant film suitable at every step in your process
- Stocked assemblies ready to ship
Related Product Literature
Specification Sheet: Mobius® Essential Assemblies
Specification Sheet: PureFlex™ Films
Flexible, robust, and chemical-resistant film for anywhere in your process train
PureFlex™ film is a high-purity, medical grade, coextruded film designed with low gas permeability and an inert product contact layer. Suited at every step in the drug manufacturing process, it enables biomanufacturers to scale their process with a single fluid-contact material. PureFlex™ film provides a fluid contact layer made of ultra-low-density polyethylene (ULDPE), a gas barrier layer made of polyethylene vinyl alcohol copolymers (EVOH), an ethylene vinyl acetate (EVA) layer helping maintain flexibility, and an outer layer made of ULDPE resin.
Mobius® Essential Tubing Assemblies