Merck
HomeCampaignsSample Program: mPAGE® TurboMix Bis-Tris Polyacrylamide Gel Casting Kits
Sample Program: mPAGE® TurboMix Bis-Tris Polyacrylamide Gel Casting Kits
mPAGE® TurboMix Bis-Tris Polyacrylamide Gel Casting Kits deliver a rapid and reproducible workflow for hand-cast protein gels. Bis-tris gel chemistry has a similar molecular weight range to standard tris-glycine, but provides multiple benefits: faster run time, sharper protein bands and longer shelf life. Learn More
Move your research forward by evaluating novel, updated electrophoresis products that deliver improved results in less time. Please complete and submit the form below.
Fields indicated by an * are required.