HPLC Analysis of Tryptic Digest of Protein Mixture on BIOshell A160 2.7 μm Peptide C18
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Peptide ES-C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 2.1 mmCONDITIONS
column
BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase
[A] 0.1% formic acid in water; [B] 0.4% formic acid in 25:75 (v/v) water:acetonitrile
gradient
6 to 70% B in 40 min
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI(+), TOF
injection
2 μL
sample
10 pmol/L in 0.1% formic acid
Description
Legal Information
null