Merck
HPLC Analysis of Glutamine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Glutamine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
standard
Product No.
Description
Pricing
L-Glutamine
meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
L-Glutamine
ReagentPlus®, ≥99% (HPLC)
L-Glutamine
γ-irradiated, BioXtra, suitable for cell culture
D-Glutamine
≥98% (HPLC)
L-Glutamine solution
200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
L-Glutamine
L-Glutamine
BioUltra, ≥99.5% (NT)
L-Glutamine
L-Glutamine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] methanol; [C] formic acid; (30:70:0.02, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
300 μg /mL in 30:70 water:methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC