HPLC Analysis of FMOC-Proline Enantiomers on Astec® CYCLOBOND I 2000 SN
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000 SN, 25 cm x 4.6 mm I.D., 5 μm particles (20524AST)
mobile phase
[A] acetonitrile; [B] methanol; [C] acetic acid; [D] ammonium hydroxide; (85:15:0.4:0.1, A:B:C:D)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null