GC Analysis of Trichloroanisole (TCA) in Wine on SLB®-5ms after SPME using 100 μm PDMS Fiber
Materials
analytical column
Product No.
Description
Pricing
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosamplerstandard
Product No.
Description
Pricing
CONDITIONS
sample/matrix
red wine spiked with 5 ppt TCA
SPME fiber
100 μm PDMS metal fiber (57928-U)
extraction
headspace, room temp (30 min)
condition
3 min. at 250 °C
desorption process
250 °C for 3 min
column
SLB®-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
60 °C (2 min), 8 °C/min to 200 °C (10 min)
MSD interface
300 °C
scan range
SIM, m/z = 195, 197, 210, 212
carrier gas
helium, 1 mL/min. constant
liner
0.75 mm I.D. SPME
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany