Merck
  • Utilization of Lactobacillus sp. for steroid glycoalkaloids degradation by lactic acid fermentation.
Utilization of Lactobacillus sp. for steroid glycoalkaloids degradation by lactic acid fermentation.
Die Nahrung (2002-09-13)
Maria Veselá, M Drdák, P Simon, M Veselý
ABSTRACT
The degradation fo steroid glycoalkaloids (SGAs) has been studied in model solutions. The number of colony forming units (CFU) was determined using a nondirect (cultivation) method during all stages of fermentation. The changes in SGAs content were observed by HPLC on the Supelcosil LC-NH2 column. The changes in alpha-tomatine concentration added to fermented Lactobacillus MRS broth have been studied. A mathematical model of steroid glycoalkaloids degradation during lactic fermentation was proposed. The mathematical model was based on the experimental data of SGA and glucose concentration and should be used for study and prediction of SGA concentration changes of fermented samples. The ratio of SGA degradation rate by fermentation and by lactic acid hydrolysis was calculated. The experimental data evaluated by proposed mathematical model for the selected strain of Lactobacillus plantarum 976H show real feasibility for SGA degradation by lactic acid fermentation.
Product Description
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
SUPELCOSIL LC-NH2 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm