Merck
  • Effect of heavy atoms on photochemically induced dynamic nuclear polarization in liquids.
Effect of heavy atoms on photochemically induced dynamic nuclear polarization in liquids.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2017-12-24)
Yusuke Okuno, Silvia Cavagnero
ABSTRACT
Given its short hyperpolarization time (∼10
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
3-Chloro-1-propanol, 98%
Sigma-Aldrich
Iodoethane, contains copper as stabilizer, ReagentPlus®, 99%
Millipore
Campylobacter Selective Supplement (Karmali), suitable for microbiology
Sigma-Aldrich
3-Bromo-1-propanol, 97%