Merck
Evaluation of carboxamide-type synthetic cannabinoids as CB
Forensic toxicology (2018-01-26)
Takahiro Doi, Takaomi Tagami, Akihiro Takeda, Akiko Asada, Yoshiyuki Sawabe
ABSTRACT
Recently, carboxamide-type synthetic cannabinoids have been distributed globally as new psychoactive substances (NPS). Some of these compounds possess asymmetric carbon, which is derived from an amide moiety composed of amino acid derivatives (i.e., amides or esters of amino acids). In this study, we synthesized both enantiomers of synthetic cannabinoids,
Sigma-Aldrich
(Bromomethyl)cyclohexane, 99%
Pricing and availability is not currently available.