Merck
  • Amyloid Precursor Proteins Are Dynamically Trafficked and Processed during Neuronal Development.
Amyloid Precursor Proteins Are Dynamically Trafficked and Processed during Neuronal Development.
Frontiers in molecular neuroscience (2016-12-10)
Jenna M Ramaker, Robert S Cargill, Tracy L Swanson, Hanil Quirindongo, Marlène Cassar, Doris Kretzschmar, Philip F Copenhaver
ABSTRACT
Proteolytic processing of the Amyloid Precursor Protein (APP) produces beta-amyloid (Aβ) peptide fragments that accumulate in Alzheimer's Disease (AD), but APP may also regulate multiple aspects of neuronal development, albeit via mechanisms that are not well understood. APP is a member of a family of transmembrane glycoproteins expressed by all higher organisms, including two mammalian orthologs (APLP1 and APLP2) that have complicated investigations into the specific activities of APP. By comparison, insects express only a single APP-related protein (APP-Like, or APPL) that contains the same protein interaction domains identified in APP. However, unlike its mammalian orthologs, APPL is only expressed by neurons, greatly simplifying an analysis of its functions
Sigma-Aldrich
Anti-APP (Ab-668) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC₅₀ = 15 nM for human BACE-1).
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).
Sigma-Aldrich
Amyloid Precursor Protein β-Secretase Inhibitor, The Amyloid Precursor Protein β-Secretase Inhibitor controls the biological activity of Amyloid Precursor Protein β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.