Merck
  • Activation of TPA-response element present in human Lemur Tyrosine Kinase 2 (
Activation of TPA-response element present in human Lemur Tyrosine Kinase 2 (
Biochemistry and biophysics reports (2017-11-02)
Isha Dey, Neil A Bradbury
ABSTRACT
Regulatory elements present in the promoter of a gene drive the expression of the gene in response to various stimuli. Lemur Tyrosine Kinase 2 (LMTK2) is a membrane-anchored Serine/Threonine kinase involved in endosomal protein trafficking and androgen signaling amongst other processes. Previous studies have shown this protein to be of therapeutic importance in cystic fibrosis and prostate cancer. However, nothing is known about the endogenous expression of this protein and its regulation. In this study, we analyzed the gene encoding human LMTK2, to look for possible regulatory elements that could affect its expression. Interestingly, the human
Sigma-Aldrich
Phorbol-12-myristate-13-acetate, Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.
Sigma-Aldrich
Anti-LMTK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution